Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
Unity Lab Services
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other B3GALNT1 SIRNA Assays ›
Gene Symbol:
B3GALNT1
Gene Name:
beta-1,3-N-acetylgalactosaminyltransferase 1 (globoside blood group)
Gene Aliases:
B3GALT3, GLCT3, GLOB, Gb4Cer, P, P1, beta3Gal-T3, galT3
Chromosome Location:
Chr.3: 161083883 - 161105528 on Build GRCh38
UniGene ID:
Hs.418062
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s16582
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: B3GALNT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001038628.1
NP_001033717.1
8
1394
NM_003781.3
NP_003772.1
5
1071
NM_033167.2
NP_149357.1
6
1190
NM_033168.2
NP_149358.1
4
976
NM_033169.2
NP_149359.1
5
1044
XM_005247859.4
XP_005247916.1
6
1134
XM_005247861.3
XP_005247918.1
7
1254
XM_005247862.3
XP_005247919.1
6
1112
XM_005247864.3
XP_005247921.1
5
1069
XM_006713803.3
XP_006713866.1
7
1253
XM_006713804.3
XP_006713867.1
4
1194
XM_006713805.3
XP_006713868.1
5
1313
XM_017007402.1
XP_016862891.1
6
1208
XM_017007403.1
XP_016862892.1
6
1156
XM_017007404.1
XP_016862893.1
6
1164
XM_017007405.1
XP_016862894.1
5
1037
XM_017007406.1
XP_016862895.1
7
1231
XM_017007407.1
XP_016862896.1
7
1380
XM_017007408.1
XP_016862897.1
6
1283
XM_017007409.1
XP_016862898.1
5
1176
XM_017007410.1
XP_016862899.1
7
1263
XM_017007411.1
XP_016862900.1
5
1185
XM_017007412.1
XP_016862901.1
4
1110
GenBank mRNA
AB050855.1
-
1
733
AB050856.1
-
5
987
AB060691.1
-
1
402
AF132731.1
-
2
700
AF154848.1
-
5
1020
AK022491.1
-
5
1023
AK223439.1
-
5
984
AY359049.1
-
4
946
BC028571.1
-
8
1316
4838965
BC047618.1
-
5
966
5259171
CR457170.1
-
1
625
Y15062.1
-
1
625
Back To Top
More Information
Panther Classification:
Molecular Function -
glycosyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
protein glycosylation
oligosaccharide biosynthetic process
UDP-galactose:beta-N-acetylglucosamine beta-1,3-galactosyltransferase activity
galactosylgalactosylglucosylceramide beta-D-acetylgalactosaminyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0