Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RAB11A SIRNA Assays ›
          Gene Symbol:
          RAB11A
          Gene Name:
          RAB11A, member RAS oncogene family
          Gene Aliases:
          YL8
          Chromosome Location:
          Chr.15: 65869459 - 65891991 on Build GRCh38
          UniGene ID:
          Hs.321541
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s16703
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 7,898.00
          Your Price
          Online offer:
          7898.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RAB11A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_004663.4 NP_004654.1 4, 5 646
          GenBank mRNA AF000231.1 - 4, 5 586
          AF498946.1 - 4, 5 519
          AK297549.1 - 4, 5 657
          AK298961.1 - 4, 5 439
          AK311770.1 - 4, 5 630
          BC013348.2 - 4, 5 613 3510339
          BI550703.1 - 4, 5 656
          BT020151.1 - 4, 5 519
          BT020154.1 - 4, 5 519
          CR407669.1 - 4, 5 519
          CR536493.1 - 4, 5 519
          X53143.1 - 4, 5 519
          X56740.1 - 4, 5 572

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00366449_g1

          Gene Ontology Categories:

          Function(s) Process(es)

          cytokinesis
          renal water homeostasis
          mitotic metaphase plate congression
          small GTPase mediated signal transduction
          positive regulation of epithelial cell migration
          regulation of multivesicular body size
          positive regulation of G2/M transition of mitotic cell cycle
          protein transport
          vesicle-mediated transport
          astral microtubule organization
          neuron projection development
          melanosome transport
          multivesicular body assembly
          positive regulation of axon extension
          regulation of long-term neuronal synaptic plasticity
          plasma membrane to endosome transport
          regulation of protein transport
          establishment of vesicle localization
          regulation of vesicle-mediated transport
          establishment of protein localization to organelle
          protein localization to plasma membrane
          establishment of protein localization to membrane
          mitotic spindle assembly
          exosomal secretion
          GTPase activity
          protein binding
          GTP binding
          microtubule binding
          syntaxin binding
          myosin V binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2zckh:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0