Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MAP3K6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297609.1 | NP_001284538.1 | 26 | 3931 | ||
NM_004672.4 | NP_004663.3 | 27 | 3955 | |||
XM_017002771.1 | XP_016858260.1 | 25 | 3336 | |||
GenBank mRNA | AB167411.1 | - | 27 | 3686 | ||
AB208805.1 | - | 26 | 3400 | |||
AF100318.1 | - | 24 | 3100 | |||
BC129950.1 | - | 19 | 2385 | |||
BC129951.1 | - | 19 | 2385 |