Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CLDN10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160100.1 | NP_001153572.1 | 3 | 581 | ||
NM_006984.4 | NP_008915.1 | 3 | 470 | |||
NM_182848.3 | NP_878268.1 | 3 | 638 | |||
XM_011521134.2 | XP_011519436.1 | 4 | 563 | |||
XM_017020843.1 | XP_016876332.1 | 4 | 775 | |||
XM_017020844.1 | XP_016876333.1 | 4 | 738 | |||
GenBank mRNA | AK055855.1 | - | 3 | 633 | ||
AK315737.1 | - | 3 | 422 | |||
BC010920.1 | - | 3 | 442 | 4246806 | ||
CR456845.1 | - | 3 | 409 | |||
DA133712.1 | - | 3 | 470 | |||
U89916.1 | - | 3 | 433 |