Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ADIPOQ SIRNA Assays ›
          Gene Symbol:
          ADIPOQ
          Gene Name:
          adiponectin, C1Q and collagen domain containing
          Gene Aliases:
          ACDC, ACRP30, ADIPQTL1, ADPN, APM-1, APM1, GBP28
          Chromosome Location:
          Chr.3: 186842674 - 186858463 on Build GRCh38
          UniGene ID:
          Hs.80485
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s17927

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ADIPOQ
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001177800.1 NP_001171271.1 4 521
          NM_004797.3 NP_004788.1 3 470
          GenBank mRNA AK291525.1 - 3 443
          AK312868.1 - 4 491
          AX767959.1 - 3 412
          BC054496.1 - 411 6192794
          BC096308.3 - 3 454 40007540
          BC096309.3 - 3 454 40007541
          BC096310.3 - 3 454 40007542
          BC096311.3 - 3 454 40007544
          D45371.1 - 3 412
          EU420013.1 - 2 386

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          positive regulation of protein phosphorylation
          glucose metabolic process
          generation of precursor metabolites and energy
          fatty acid beta-oxidation
          response to nutrient
          circadian rhythm
          response to sucrose
          response to glucose
          positive regulation of signal transduction
          negative regulation of platelet-derived growth factor receptor signaling pathway
          positive regulation of protein kinase A signaling
          negative regulation of macrophage derived foam cell differentiation
          negative regulation of tumor necrosis factor-mediated signaling pathway
          positive regulation of cholesterol efflux
          regulation of glucose metabolic process
          response to activity
          negative regulation of smooth muscle cell migration
          fatty acid oxidation
          negative regulation of cell migration
          negative regulation of granulocyte differentiation
          negative regulation of protein autophosphorylation
          positive regulation of cellular protein metabolic process
          negative regulation of tumor necrosis factor production
          positive regulation of interleukin-8 production
          cellular response to insulin stimulus
          positive regulation of myeloid cell apoptotic process
          adiponectin-activated signaling pathway
          negative regulation of heterotypic cell-cell adhesion
          low-density lipoprotein particle clearance
          response to tumor necrosis factor
          cellular response to drug
          glucose homeostasis
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of MAP kinase activity
          response to ethanol
          negative regulation of fat cell differentiation
          negative regulation of macrophage differentiation
          negative regulation of low-density lipoprotein particle receptor biosynthetic process
          negative regulation of gluconeogenesis
          negative regulation of blood pressure
          positive regulation of blood pressure
          negative regulation of transcription, DNA-templated
          positive regulation of fatty acid metabolic process
          positive regulation of glucose import
          negative regulation of hormone secretion
          negative regulation of smooth muscle cell proliferation
          negative regulation of inflammatory response
          positive regulation of peptidyl-tyrosine phosphorylation
          negative regulation of phagocytosis
          negative regulation of synaptic transmission
          brown fat cell differentiation
          protein homooligomerization
          response to glucocorticoid
          membrane depolarization
          membrane hyperpolarization
          protein heterotrimerization
          negative regulation of ERK1 and ERK2 cascade
          response to linoleic acid
          detection of oxidative stress
          cellular response to cAMP
          positive regulation of monocyte chemotactic protein-1 production
          cellular response to epinephrine stimulus
          protein localization to plasma membrane
          negative regulation of intracellular protein transport
          negative regulation of receptor binding
          negative regulation of DNA biosynthetic process
          positive regulation of glycogen (starch) synthase activity
          positive regulation of metanephric glomerular visceral epithelial cell development
          positive regulation of cAMP-dependent protein kinase activity
          positive regulation of renal albumin absorption
          negative regulation of platelet-derived growth factor receptor-alpha signaling pathway
          negative regulation of metanephric mesenchymal cell migration
          receptor binding
          cytokine activity
          hormone activity
          protein binding
          sialic acid binding
          identical protein binding
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0