Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: DLK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286656.1 | NP_001273585.1 | 5 | 617 | ||
NM_023932.3 | NP_076421.2 | 5 | 635 | |||
NM_206539.2 | NP_996262.1 | 5 | 1051 | |||
XM_005249308.4 | XP_005249365.1 | 4 | 1843 | |||
XM_017011204.1 | XP_016866693.1 | 2 | 653 | |||
GenBank mRNA | AK055380.1 | - | 5 | 1051 | ||
AY358126.1 | - | 5 | 501 | |||
BC000230.1 | - | 5 | 467 | 3350499 | ||
BC006425.1 | - | 5 | 467 | 3349698 | ||
BC110320.1 | - | 5 | 603 | 5495458 | ||
BI916137.1 | - | 5 | 559 | |||
BM811564.1 | - | 582 |