Search
Search

Kcnk6| Gene Symbol: Kcnk6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033525.3 | NP_001028697.2 | 2 | 580 | ||
| GenBank mRNA | AK036882.1 | - | 2 | 535 | ||
| AK132480.1 | - | 2 | 581 | |||
| AK163847.1 | - | 2 | 429 | |||
| AK171038.1 | - | 2 | 574 | |||
| BC115791.1 | - | 2 | 286 | 40100611 | ||
| BC115915.1 | - | 2 | 280 | 40100605 | ||
| DQ185135.1 | - | 2 | 541 | |||