Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LILRB3 SIRNA Assays ›
Gene Symbol:
LILRB3
LILRA6
Gene Name:
leukocyte immunoglobulin like receptor B3
leukocyte immunoglobulin like receptor A6
Gene Aliases:
CD85A, HL9, ILT-5, ILT5, LILRA6, LIR-3, LIR3, PIR-B, PIRB
CD85b, ILT-8, ILT5, ILT8, LILRB3, LILRB6
Chromosome Location:
Chr.19: 54216278 - 54223087 on Build GRCh38
Chromosome Location:
Chr.19: 54236592 - 54242768 on Build GRCh38
UniGene ID:
Hs.631592
Hs.688335
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
s21720
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LILRB3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001081450.2
NP_001074919.2
8
1434
NM_001320960.1
NP_001307889.1
9
1485
NM_006864.3
NP_006855.3
8
1434
NR_135493.1
8
1434
NR_135494.1
8
1348
NR_135495.1
8
1354
NR_135496.1
9
1517
XM_011526381.2
XP_011524683.1
9
1485
XM_011526382.2
XP_011524684.1
9
1485
XM_017026218.1
XP_016881707.1
8
1353
XM_017026219.1
XP_016881708.1
8
1353
XM_017026220.1
XP_016881709.1
8
1353
XM_017026221.1
XP_016881710.1
8
1353
XM_017026222.1
XP_016881711.1
7
1302
XM_017026223.1
XP_016881712.1
7
1302
GenBank mRNA
AF000575.1
-
8
1323
AF009632.1
-
8
1325
AF009633.1
-
8
1325
AF009634.1
-
8
1325
AF009635.1
-
8
1325
AF009636.1
-
8
1325
AF009637.1
-
9
1376
AF009638.1
-
8
1325
AF009639.1
-
8
1325
AF009640.1
-
8
1325
AF009641.1
-
8
1325
AF009642.1
-
8
1325
AF009643.1
-
8
1239
AF009644.1
-
8
1245
AF025533.1
-
8
1389
AF031553.1
-
8
1325
AF031554.1
-
8
1325
AF031555.1
-
8
1325
AF031556.1
-
8
1325
AK223265.1
-
8
1376
AK290917.1
-
8
1404
BC028153.1
-
9
1408
BC104993.1
-
8
1342
8143996
BC112198.1
-
8
1342
8327666
EU814627.1
-
8
1323
EU814628.1
-
8
1323
KC918852.1
-
8
1367
KC918854.1
-
8
1367
KC918855.1
-
8
1367
KC918857.1
-
8
1367
KC918858.1
-
8
1367
KC918860.1
-
8
1367
KC918863.1
-
8
1367
U91928.1
-
8
1372
Gene Symbol: LILRA6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
GenBank mRNA
KC918856.1
-
8
1367
KC918859.1
-
9
1367
Back To Top
More Information
Panther Classification:
Molecular Function -
immunoglobulin receptor superfamily
Gene Ontology Categories:
Function(s)
Process(es)
adaptive immune response
defense response
cell surface receptor signaling pathway
negative regulation of osteoclast differentiation
receptor activity
transmembrane signaling receptor activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-l5829:80/100.66.79.204:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0