Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FTL1 SIRNA Assays ›
          Gene Symbol:
          Ftl1 LOC100360087
          Gene Name:
          ferritin light chain 1
          ferritin light chain 1-like
          Gene Aliases:
          Ftl
          -
          Chromosome Location:
          Chr.1: 101448190 - 101450034 on Build Rnor_6.0
          Chromosome Location:
          Chr.5: 152325856 - 152326797 on Build Rnor_6.0
          UniGene ID:
          Rn.1905
          -
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s218073

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ftl1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_022500.4 NP_071945.3 4 856
          GenBank mRNA BC061525.1 - 4 860 6921603
          BC086583.1 - 4 837 7307718
          BC088756.1 - 4 837 7307747
          FQ209407.1 - 4 856
          FQ209694.1 - 4 856
          FQ209816.1 - 4 856
          FQ218577.1 - 4 856
          FQ219483.1 - 4 856
          FQ219809.1 - 4 856
          FQ219930.1 - 4 855
          FQ220095.1 - 4 856
          FQ220123.1 - 4 856
          FQ220243.1 - 4 853
          FQ220651.1 - 4 856
          FQ220814.1 - 4 856
          FQ220884.1 - 4 856
          FQ221898.1 - 4 857
          FQ222411.1 - 3 760
          FQ223227.1 - 4 856
          FQ226254.1 - 4 812
          FQ226458.1 - 4 855
          FQ226986.1 - 4 853
          FQ227401.1 - 4 856
          FQ227800.1 - 4 856
          FQ227843.1 - 4 858
          FQ228050.1 - 4 856
          FQ228585.1 - 4 856
          FQ229176.1 - 4 856
          FQ229284.1 - 4 856
          FQ229370.1 - 4 856
          FQ229673.1 - 4 854
          FQ229757.1 - 4 856
          FQ230098.1 - 4 854
          FQ230445.1 - 4 856
          FQ230659.1 - 4 852
          FQ231322.1 - 4 856
          FQ231373.1 - 4 856
          FQ231415.1 - 3 712
          FQ231464.1 - 4 856
          FQ231570.1 - 4 856
          FQ231615.1 - 4 856
          FQ231781.1 - 4 852
          FQ231957.1 - 4 856
          FQ232031.1 - 4 856
          FQ232167.1 - 4 856
          FQ232317.1 - 4 812
          FQ233996.1 - 4 856
          FQ234074.1 - 4 856
          FQ234647.1 - 4 856
          FQ235250.1 - 4 856
          K01930.1 - 4 781
          L01122.1 - 4 809
          Gene Symbol: LOC100360087
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_002729553.4 XP_002729599.1 1 871

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_010240, BC019840, AK145265, AK152385, BC106146, and others.

          Panther Classification:

          Molecular Function -

          storage protein

          Gene Ontology Categories:

          Function(s) Process(es)

          iron ion transport
          cellular iron ion homeostasis
          response to lead ion
          molecular_function
          iron ion binding
          ferric iron binding
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0