Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ZNF655 SIRNA Assays ›
          Gene Symbol:
          ZNF655 LOC100131257
          Gene Name:
          zinc finger protein 655
          zinc finger protein 655 pseudogene
          Gene Aliases:
          VIK, VIK-1
          -
          Chromosome Location:
          Chr.7: 99557726 - 99576453 on Build GRCh38
          Chromosome Location:
          Chr.7: 7075770 - 7096786 on Build GRCh38
          UniGene ID:
          Hs.599798
          Hs.551110
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s224807

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ZNF655
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001009960.1 NP_001009960.1 3 1312
          NM_001083956.1 NP_001077425.1 4 1590
          NM_001085368.1 NP_001078837.1 4 1417
          NM_138494.2 NP_612503.1 3 1485
          XM_017012598.1 XP_016868087.1 6 2490
          XM_017012599.1 XP_016868088.1 6 1648
          XM_017012600.1 XP_016868089.1 6 1661
          XM_017012601.1 XP_016868090.1 6 1647
          XM_017012602.1 XP_016868091.1 5 2385
          XM_017012603.1 XP_016868092.1 4 2510
          XM_017012604.1 XP_016868093.1 4 1427
          XM_017012605.1 XP_016868094.1 5 1438
          XM_017012606.1 XP_016868095.1 3 1343
          XM_017012607.1 XP_016868096.1 3 1329
          XM_017012608.1 XP_016868097.1 6 2523
          XM_017012609.1 XP_016868098.1 5 1537
          XM_017012610.1 XP_016868099.1 5 1484
          GenBank mRNA AK122929.1 - 1 569
          AK128057.1 - 2 2549
          AK304785.1 - 4 1294
          AK314593.1 - 3 1328
          AK316024.1 - 4 1408
          AY099353.1 - 3 1311
          BC011816.1 - 1 580
          BC024770.1 - 1 831
          BC037407.1 - 3 1275 5752559
          CR749270.1 - 3 1093
          Gene Symbol: LOC100131257
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_034022.1 1 18111

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          negative regulation of G1/S transition of mitotic cell cycle
          molecular_function
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0