Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CAMK2G SIRNA Assays ›
          Gene Symbol:
          CAMK2G
          Gene Name:
          calcium/calmodulin dependent protein kinase II gamma
          Gene Aliases:
          CAMK, CAMK-II, CAMKG
          Chromosome Location:
          Chr.10: 73812501 - 73874591 on Build GRCh38
          UniGene ID:
          Hs.523045
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s224890

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CAMK2G
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001204492.1 NP_001191421.1 8 699
          NM_001222.3 NP_001213.2 8 699
          NM_001320898.1 NP_001307827.1 8 699
          NM_172169.2 NP_751909.1 8 699
          NM_172170.4 NP_751910.1 8 699
          NM_172171.2 NP_751911.1 8 699
          NM_172173.2 NP_751913.1 8 699
          XM_005270195.1 XP_005270252.1 8 690
          XM_005270197.1 XP_005270254.1 8 690
          XM_005270198.1 XP_005270255.1 8 690
          XM_005270199.1 XP_005270256.1 8 690
          XM_005270200.1 XP_005270257.1 8 690
          XM_005270201.1 XP_005270258.1 8 690
          XM_005270203.1 XP_005270260.1 8 690
          XM_005270205.1 XP_005270262.1 8 690
          XM_006717993.1 XP_006718056.1 8 690
          XM_006717996.1 XP_006718059.1 8 690
          XM_006717997.1 XP_006718060.1 8 690
          XM_006717998.1 XP_006718061.1 8 690
          XM_011540225.2 XP_011538527.1 8 638
          XM_017016716.1 XP_016872205.1 8 690
          XM_017016717.1 XP_016872206.1 8 638
          XM_017016718.1 XP_016872207.1 8 690
          XM_017016719.1 XP_016872208.1 8 690
          XM_017016720.1 XP_016872209.1 8 690
          XM_017016721.1 XP_016872210.1 8 765
          XM_017016722.1 XP_016872211.1 8 690
          XM_017016723.1 XP_016872212.1 8 690
          XM_017016724.1 XP_016872213.1 8 690
          XM_017016725.1 XP_016872214.1 8 690
          XM_017016726.1 XP_016872215.1 8 690
          XM_017016727.1 XP_016872216.1 8 690
          XM_017016728.1 XP_016872217.1 8 638
          XM_017016729.1 XP_016872218.1 6 425
          XM_017016730.1 XP_016872219.1 8 690
          XM_017016731.1 XP_016872220.1 7 596
          XM_017016732.1 XP_016872221.1 6 463
          XM_017016733.1 XP_016872222.1 8 638
          XM_017016734.1 XP_016872223.1 7 622
          XM_017016735.1 XP_016872224.1 8 690
          XM_017016736.1 XP_016872225.1 8 765
          XM_017016737.1 XP_016872226.1 7 592
          XM_017016738.1 XP_016872227.1 8 651
          XM_017016739.1 XP_016872228.1 8 822
          GenBank mRNA AK095576.1 - 7 675
          AK131199.1 - 8 622
          AK225771.1 - 8 699
          AK291704.1 - 8 691
          BC021269.2 - 6 374
          BC034044.1 - 8 690 4813948
          BU165166.1 - 8 697
          L07043.1 - 6 374
          L07044.1 - 8 590
          U66064.1 - 8 622

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_133605, XM_008770499, XM_008770497, XM_008770639, XM_008770495, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001039139, NM_001039138, NM_178597, AK154764, BC019162, and others.

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          MAPK cascade
          protein dephosphorylation
          calcium ion transport
          nervous system development
          regulation of skeletal muscle adaptation
          insulin secretion
          cell differentiation
          positive regulation of GTPase activity
          protein autophosphorylation
          protein oligomerization
          regulation of calcium ion transport
          interferon-gamma-mediated signaling pathway
          regulation of cellular response to heat
          regulation of relaxation of cardiac muscle
          protein serine/threonine kinase activity
          calmodulin-dependent protein kinase activity
          calcium-dependent protein serine/threonine phosphatase activity
          Ras guanyl-nucleotide exchange factor activity
          protein binding
          calmodulin binding
          ATP binding
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0