Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NLGN1 SIRNA Assays ›
Gene Symbol:
NLGN1
Gene Name:
neuroligin 1
Gene Aliases:
NL1
Chromosome Location:
Chr.3: 173395899 - 174291287 on Build GRCh38
UniGene ID:
Hs.478289
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s22548
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NLGN1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_014932.3
NP_055747.1
5, 6
1296
XM_005247234.2
XP_005247291.1
4, 5
1471
XM_005247235.3
XP_005247292.1
4, 5
1667
XM_005247236.2
XP_005247293.1
4, 5
1444
XM_005247237.3
XP_005247294.1
3, 4
1965
XM_006713540.3
XP_006713603.1
5, 6
1727
XM_011512551.2
XP_011510853.1
6, 7
1578
XM_011512552.2
XP_011510854.1
5, 6
2014
XM_011512553.2
XP_011510855.1
2, 3
367
XM_011512554.2
XP_011510856.1
2, 3
327
XM_017005888.1
XP_016861377.1
5, 6
1875
XM_017005889.1
XP_016861378.1
5, 6
1875
XM_017005890.1
XP_016861379.1
5, 6
1531
XM_017005891.1
XP_016861380.1
5, 6
1727
XM_017005892.1
XP_016861381.1
5, 6
2029
XM_017005893.1
XP_016861382.1
6, 7
2141
XM_017005894.1
XP_016861383.1
5, 6
3659
XM_017005895.1
XP_016861384.1
5, 6
7832
XM_017005896.1
XP_016861385.1
5, 6
1313
XM_017005897.1
XP_016861386.1
5, 6
1848
XM_017005898.1
XP_016861387.1
4, 5
1815
XM_017005899.1
XP_016861388.1
4, 5
1954
XM_017005900.1
XP_016861389.1
5, 6
1460
XM_017005901.1
XP_016861390.1
4, 5
1516
XM_017005902.1
XP_016861391.1
5, 6
1491
XM_017005903.1
XP_016861392.1
4, 5
1640
XM_017005904.1
XP_016861393.1
4, 5
1927
XM_017005905.1
XP_016861394.1
2, 3
1658
GenBank mRNA
AB028993.1
-
4, 5
1536
AK307813.1
-
6, 7
1422
BC032555.1
-
5, 6
1301
5558890
GQ489206.1
-
5, 6
987
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_053868, XM_008760890, U22952.
This siRNA also targets the following transcripts in Mouse: NM_001163387, NM_138666, BC098461, AK122433, AK138209.
Panther Classification:
Molecular Function -
cell adhesion molecule
Gene Ontology Categories:
Function(s)
Process(es)
regulation of respiratory gaseous exchange by neurological system process
protein targeting
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
neuron cell-cell adhesion
nervous system development
synapse assembly
positive regulation of circadian sleep/wake cycle, wakefulness
synaptic vesicle targeting
calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
neuronal signal transduction
neuron projection development
positive regulation of synaptic transmission, GABAergic
protein localization to synapse
establishment of protein localization
regulation of neuron differentiation
rhythmic process
cytoskeletal matrix organization at active zone
protein homooligomerization
protein heterotetramerization
positive regulation of filopodium assembly
positive regulation of synapse assembly
positive regulation of synaptic transmission, glutamatergic
long-term synaptic potentiation
positive regulation of dendritic spine development
negative regulation of dendritic spine morphogenesis
terminal button organization
synaptic vesicle clustering
postsynaptic membrane assembly
presynaptic membrane assembly
AMPA glutamate receptor clustering
NMDA glutamate receptor clustering
neurexin clustering involved in presynaptic membrane assembly
postsynaptic density protein 95 clustering
receptor localization to synapse
positive regulation of ruffle assembly
positive regulation of synaptic vesicle endocytosis
positive regulation of protein localization to synapse
positive regulation of intracellular signal transduction
positive regulation of synaptic vesicle exocytosis
regulation of N-methyl-D-aspartate selective glutamate receptor activity
regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity
positive regulation of excitatory postsynaptic potential
positive regulation of synaptic vesicle clustering
receptor activity
PDZ domain binding
neurexin family protein binding
protein dimerization activity
cell adhesion molecule binding
carboxylic ester hydrolase activity
scaffold protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0