Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CCT5 SIRNA Assays ›
          Gene Symbol:
          CCT5
          Gene Name:
          chaperonin containing TCP1 subunit 5
          Gene Aliases:
          CCT-epsilon, CCTE, HEL-S-69, PNAS-102, TCP-1-epsilon
          Chromosome Location:
          Chr.5: 10249921 - 10266412 on Build GRCh38
          UniGene ID:
          Hs.1600
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s22729

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CCT5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001306153.1 NP_001293082.1 11 1534
          NM_001306154.1 NP_001293083.1 10 1769
          NM_001306155.1 NP_001293084.1 10 1601
          NM_001306156.1 NP_001293085.1 11 1766
          NM_012073.4 NP_036205.1 11 1934
          GenBank mRNA AF275798.1 - 5 542
          AJ012489.1 - 1 43
          AK289353.1 - 11 1532
          AK290393.1 - 11 1575
          AK293341.1 - 11 1603
          AK293437.1 - 11 1475
          AK301760.1 - 11 1526
          AK301985.1 - 12 1492
          AK302368.1 - 10 1474
          AK302383.1 - 10 1599
          AK303086.1 - 12 1461
          AK316376.1 - 11 1521
          BC002971.1 - 11 1509
          BC006543.2 - 11 1549 2824199
          BC009454.1 - 8 1053
          BC035499.2 - 11 1596 4329194
          BC040027.1 - 11 1566
          D43950.2 - 11 1551
          FJ224303.1 - 11 1685

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          chaperonin

          Gene Ontology Categories:

          Function(s) Process(es)

          protein folding
          binding of sperm to zona pellucida
          response to virus
          positive regulation of telomere maintenance via telomerase
          protein stabilization
          toxin transport
          positive regulation of establishment of protein localization to telomere
          positive regulation of protein localization to Cajal body
          positive regulation of telomerase RNA localization to Cajal body
          protein binding
          ATP binding
          G-protein beta-subunit binding
          beta-tubulin binding
          unfolded protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0