Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NFASC SIRNA Assays ›
          Gene Symbol:
          NFASC
          Gene Name:
          neurofascin
          Gene Aliases:
          NF, NRCAML
          Chromosome Location:
          Chr.1: 204828652 - 205022822 on Build GRCh38
          UniGene ID:
          Hs.13349
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s23064

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NFASC
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001005388.2 NP_001005388.2 28, 29 3746
          NM_001160331.1 NP_001153803.1 25, 26 3355
          NM_001160332.1 NP_001153804.1 26, 27 3548
          NM_015090.3 NP_055905.2 25, 26 3533
          XM_005244989.3 XP_005245046.2 27, 28 3673
          XM_005244991.3 XP_005245048.2 26, 27 3628
          XM_005244992.4 XP_005245049.2 25, 26 3613
          XM_005244993.3 XP_005245050.2 25, 26 3577
          XM_011509311.1 XP_011507613.1 28, 29 3865
          XM_011509312.1 XP_011507614.1 30, 31 4180
          XM_011509313.1 XP_011507615.1 28, 29 3835
          XM_011509314.1 XP_011507616.1 29, 30 4129
          XM_011509315.1 XP_011507617.1 27, 28 3784
          XM_011509316.1 XP_011507618.1 31, 32 4077
          XM_011509317.1 XP_011507619.1 28, 29 3727
          XM_011509318.2 XP_011507620.1 30, 31 4102
          XM_011509319.1 XP_011507621.1 30, 31 4026
          XM_011509320.2 XP_011507622.1 29, 30 4051
          XM_011509321.1 XP_011507623.1 28, 29 3643
          XM_011509322.1 XP_011507624.1 27, 28 3610
          XM_011509323.1 XP_011507625.1 27, 28 3559
          XM_011509324.1 XP_011507626.1 26, 27 3373
          XM_011509325.1 XP_011507627.1 25, 26 3358
          XM_011509326.1 XP_011507628.1 24, 25 3052
          XM_011509327.1 XP_011507629.1 23, 24 3037
          XM_011509328.1 XP_011507630.1 23, 24 3001
          XM_017000733.1 XP_016856222.1 29, 30 4135
          XM_017000734.1 XP_016856223.1 28, 29 4084
          XM_017000735.1 XP_016856224.1 27, 28 3628
          XM_017000736.1 XP_016856225.1 25, 26 3328
          XM_017000738.1 XP_016856227.1 26, 27 3550
          XM_017000739.1 XP_016856228.1 25, 26 3535
          XM_017000740.1 XP_016856229.1 23, 24 3214
          XM_017000741.1 XP_016856230.1 27, 28 3670
          XM_017000742.1 XP_016856231.1 26, 27 3495
          GenBank mRNA AB018299.3 - 24, 25 3364
          AK027553.1 - 4, 5 488
          AK127424.1 - 6, 7 828
          AK128699.1 - 4, 5 1322
          BC117674.2 - 27, 28 3595
          BC137013.1 - 23, 24 3270 9021006
          BC144454.1 - 26, 27 3483 9052980
          BC144455.1 - 25, 26 3422
          BX537841.1 - 2, 3 352
          CR749402.1 - 1, 2 1241

          Back To Top

          More Information


          Set Membership:

          G-protein coupled receptor

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          protein localization to paranode region of axon
          axon guidance
          peripheral nervous system development
          transmission of nerve impulse
          paranodal junction assembly
          heterotypic cell-cell adhesion
          myelination
          clustering of voltage-gated sodium channels
          synapse organization
          protein localization to juxtaparanode region of axon
          protein targeting to plasma membrane
          protein binding
          protein domain specific binding
          protein binding involved in heterotypic cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-4c88c:80/100.66.77.220:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0