Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Oxnad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145460.2 | NP_663435.2 | 5 | 694 | ||
GenBank mRNA | AK028649.1 | - | 5 | 604 | ||
AK031978.1 | - | 4 | 323 | |||
AK032160.1 | - | 5 | 611 | |||
AK075924.1 | - | 4 | 321 | |||
AK139884.1 | - | 5 | 694 | |||
AK215115.1 | - | 4 | 352 | |||
AK215185.1 | - | 4 | 352 | |||
BC019806.1 | - | 5 | 472 | 4018387 |