Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HIST1H4H SIRNA Assays ›
          Gene Symbol:
          Hist1h4h Hist1h4a Hist1h4b Hist1h4c Hist1h4d Hist1h4f Hist1h4i Hist1h4j Hist1h4k Hist1h4m Hist1h4n Hist2h4
          Gene Name:
          histone cluster 1, H4h
          histone cluster 1, H4a
          histone cluster 1, H4b
          histone cluster 1, H4c
          histone cluster 1, H4d
          histone cluster 1, H4f
          histone cluster 1, H4i
          histone cluster 1, H4j
          histone cluster 1, H4k
          histone cluster 1, H4m
          histone cluster 1, H4n
          histone cluster 2, H4
          Gene Aliases:
          1700024H08Rik
          -
          -
          -
          -
          -
          -
          -
          -
          Gm11275, OTTMUSG00000000446
          -
          H4, X04652
          Chromosome Location:
          Chr.13: 23531044 - 23531478 on Build GRCm38
          Chromosome Location:
          Chr.13: 23760795 - 23761249 on Build GRCm38
          Chromosome Location:
          Chr.13: 23756937 - 23757386 on Build GRCm38
          Chromosome Location:
          Chr.13: 23698084 - 23698458 on Build GRCm38
          Chromosome Location:
          Chr.13: 23581602 - 23581969 on Build GRCm38
          Chromosome Location:
          Chr.13: 23551286 - 23551643 on Build GRCm38
          Chromosome Location:
          Chr.13: 22040960 - 22041362 on Build GRCm38
          Chromosome Location:
          Chr.13: 21735096 - 21735407 on Build GRCm38
          Chromosome Location:
          Chr.13: 21750145 - 21750553 on Build GRCm38
          Chromosome Location:
          Chr.13: 21811746 - 21812150 on Build GRCm38
          Chromosome Location:
          Chr.13: 21831793 - 21832158 on Build GRCm38
          Chromosome Location:
          Chr.3: 96262934 - 96263317 on Build GRCm38
          UniGene ID:
          Mm.158272
          Mm.228709
          Mm.261642
          Mm.261662
          Mm.260530
          Mm.442307
          Mm.14775
          Mm.486099
          Mm.255646
          Mm.442307
          Mm.261664
          Mm.227295
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s234191

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Hist1h4h
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_153173.4 NP_694813.1 1 252
          GenBank mRNA AA028777.1 - 1 250
          AK007642.1 - 1 248
          BC028550.1 - 1 193 1398106
          BC058529.1 - 1 189 6817839
          BC087952.1 - 1 241 6787571
          Gene Symbol: Hist1h4a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178192.2 NP_835499.1 1 273
          GenBank mRNA AK016310.1 - 1 255
          BC119241.1 - 1 273 8733990
          BC119243.1 - 1 273 8733992
          Gene Symbol: Hist1h4b
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178193.2 NP_835500.1 1 265
          GenBank mRNA BC132186.1 - 1 265 40130463
          BC132212.1 - 1 265 40130489
          Gene Symbol: Hist1h4c
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178208.2 NP_835515.1 1 190
          GenBank mRNA BC115450.1 - 1 181 40099776
          BC115451.1 - 1 174
          BC139809.1 - 1 181 40099779
          Gene Symbol: Hist1h4d
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_175654.2 NP_783585.1 1 181
          GenBank mRNA BC119611.2 - 1 180
          Gene Symbol: Hist1h4f
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_175655.2 NP_783586.1 1 181
          GenBank mRNA BC115446.1 - 1 174
          Gene Symbol: Hist1h4i
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_175656.3 NP_783587.1 1 220
          GenBank mRNA AK011560.1 - 1 221
          BC019757.1 - 1 210
          BC052219.1 - 1 193
          BC092144.1 - 1 210 30612926
          BC125598.1 - 1 204 40130113
          BC125600.1 - 1 204 40130115
          Gene Symbol: Hist1h4j
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178210.1 NP_835582.1 1 181
          GenBank mRNA AK010085.1 - 1 217
          Gene Symbol: Hist1h4k
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178211.2 NP_835583.1 1 229
          GenBank mRNA AK139521.1 - 1 216
          BC096767.1 - 196
          BC117010.1 - 1 229 40126329
          BC117012.1 - 1 229 40126331
          BC119612.1 - 1 181 40061668
          Gene Symbol: Hist1h4m
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001195421.1 NP_001182350.1 1 219
          GenBank mRNA BC139425.1 - 243 8862447
          BC139437.1 - 243 8862459
          Gene Symbol: Hist1h4n
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_175657.2 NP_783588.1 1 181
          GenBank mRNA BC115448.2 - 1 181 40099764
          BC152397.1 - 1 181 40099768
          Gene Symbol: Hist2h4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_033596.3 NP_291074.1 1 194
          GenBank mRNA BC051566.1 - 1 202
          BC057955.1 - 188 5359141
          BX520915.1 - 1 194

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_003548, NM_001034077, BC093969, BC108260, CR542172, and others.
          This siRNA also targets the following transcripts in Rat: NM_001271221, NM_022686, XM_008771773, XM_008771675, XM_008771676, and others.

          Panther Classification:

          Molecular Function -

          histone

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleosome assembly
          DNA replication-dependent nucleosome assembly
          DNA replication-independent nucleosome assembly
          negative regulation of megakaryocyte differentiation
          protein heterotetramerization
          histone binding
          poly(A) RNA binding
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0