Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cebpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287577.1 | NP_001274506.1 | 1 | 1023 | ||
NM_001287578.1 | NP_001274507.1 | 1 | 1023 | |||
NM_001287579.1 | NP_001274508.1 | 1 | 1023 | |||
NM_012524.3 | NP_036656.1 | 1 | 1023 |