Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CAT SIRNA Assays ›
          Gene Symbol:
          CAT
          Gene Name:
          catalase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 34438925 - 34472060 on Build GRCh38
          UniGene ID:
          Hs.502302
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s2444
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 7,898.00
          Your Price
          Online offer:
          7898.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CAT
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001752.3 NP_001743.1 12, 13 1615
          GenBank mRNA AK291585.1 - 12, 13 1615
          AK315350.1 - 12, 13 1599
          AY028632.1 - 12, 13 1526
          BC027300.1 - 5, 6 629
          BC110398.1 - 12, 13 1604 5551309
          BC112217.1 - 12, 13 1564 8327685
          BC112219.1 - 12, 13 1564 8327687
          X04076.1 - 12, 13 1596

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00156308_m1

          Panther Classification:

          Molecular Function -

          peroxidase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to reactive oxygen species
          osteoblast differentiation
          ureteric bud development
          response to hypoxia
          purine nucleotide catabolic process
          triglyceride metabolic process
          aging
          cholesterol metabolic process
          aerobic respiration
          response to light intensity
          UV protection
          response to ozone
          response to lead ion
          positive regulation of phosphatidylinositol 3-kinase signaling
          response to activity
          response to inactivity
          hemoglobin metabolic process
          negative regulation of NF-kappaB transcription factor activity
          response to estradiol
          response to insulin
          response to vitamin A
          response to vitamin E
          response to L-ascorbic acid
          response to drug
          response to hydrogen peroxide
          hydrogen peroxide catabolic process
          negative regulation of apoptotic process
          response to ethanol
          response to cadmium ion
          positive regulation of NF-kappaB transcription factor activity
          protein tetramerization
          protein homotetramerization
          positive regulation of cell division
          response to hyperoxia
          response to fatty acid
          cellular response to growth factor stimulus
          response to phenylpropanoid
          cellular oxidant detoxification
          aminoacylase activity
          catalase activity
          receptor binding
          antioxidant activity
          oxidoreductase activity, acting on peroxide as acceptor
          enzyme binding
          heme binding
          protein homodimerization activity
          metal ion binding
          NADP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0