Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
Unity Lab Services
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DNM3 SIRNA Assays ›
Gene Symbol:
DNM3
Gene Name:
dynamin 3
Gene Aliases:
Dyna III
Chromosome Location:
Chr.1: 171841478 - 172418466 on Build GRCh38
UniGene ID:
Hs.654775
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s25019
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DNM3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001136127.2
NP_001129599.1
10
1467
NM_001278252.1
NP_001265181.1
10
1467
NM_015569.4
NP_056384.2
10
1467
XM_005245079.1
XP_005245136.1
10
1450
XM_005245080.1
XP_005245137.1
10
1450
XM_006711268.2
XP_006711331.1
10
1450
XM_017000976.1
XP_016856465.1
10
1450
XM_017000977.1
XP_016856466.1
10
1450
XM_017000978.1
XP_016856467.1
10
1450
XM_017000979.1
XP_016856468.1
10
1450
XM_017000980.1
XP_016856469.1
10
1168
XM_017000981.1
XP_016856470.1
10
1450
XM_017000982.1
XP_016856471.1
10
1450
XM_017000983.1
XP_016856472.1
10
1450
XM_017000984.1
XP_016856473.1
10
1450
XM_017000985.1
XP_016856474.1
10
1450
XM_017000986.1
XP_016856475.1
10
1791
XM_017000987.1
XP_016856476.1
10
1322
XM_017000988.1
XP_016856477.1
10
1450
XM_017000989.1
XP_016856478.1
10
1450
XM_017000990.1
XP_016856479.1
10
1450
XM_017000991.1
XP_016856480.1
10
1450
XM_017000992.1
XP_016856481.1
10
1450
GenBank mRNA
AB020627.2
-
10
1418
AK056279.1
-
10
1447
AK307806.1
-
10
1405
AK308992.1
-
10
1416
AL136712.1
-
10
1445
BC064546.1
-
10
1308
5744324
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
Gene Ontology Categories:
Function(s)
Process(es)
endocytosis
synapse assembly
filopodium assembly
positive regulation of filopodium assembly
negative regulation of dendritic spine morphogenesis
GTPase activity
protein binding
GTP binding
type 1 metabotropic glutamate receptor binding
type 5 metabotropic glutamate receptor binding
nitric-oxide synthase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-4d29m:80/100.66.76.134:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0