Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RRN3 SIRNA Assays ›
          Gene Symbol:
          RRN3 RRN3P1 RRN3P2
          Gene Name:
          RRN3 homolog, RNA polymerase I transcription factor
          RRN3 homolog, RNA polymerase I transcription factor pseudogene 1
          RRN3 homolog, RNA polymerase I transcription factor pseudogene 2
          Gene Aliases:
          A-270G1.2, TIFIA
          -
          -
          Chromosome Location:
          Chr.16: 15060022 - 15094335 on Build GRCh38
          Chromosome Location:
          Chr.16: 21796630 - 21819174 on Build GRCh38
          Chromosome Location:
          Chr.16: 29074842 - 29116718 on Build GRCh38
          UniGene ID:
          Hs.460078
          Hs.31290
          Hs.720348
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID s29325

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 392.00
          Your Price
          Online offer: 380.65
          380.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RRN3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001301064.1 NP_001287993.1 13 1364
          NM_018427.4 NP_060897.3 14 1454
          XM_005255376.1 XP_005255433.1 12 1229
          XM_005255377.4 XP_005255434.1 11 1178
          XM_011522540.2 XP_011520842.1 9 866
          XM_011522541.1 XP_011520843.1 13 1293
          GenBank mRNA AF227156.1 - 14 1420
          AJ272050.1 - 14 1374
          AK225662.1 - 14 1430
          AK295486.1 - 11 1047
          AK300253.1 - 13 1273
          AK302994.1 - 11 1101
          AK303405.1 - 10 947
          AK304856.1 - 14 1320
          AK314769.1 - 14 1368
          AK316520.1 - 13 1278
          BC036182.1 - 14 1369
          BC071868.1 - 6 689
          BC132688.1 - 14 1371 40146710
          BC132690.1 - 14 1371 40146712
          Gene Symbol: RRN3P1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003370.2 10 925
          GenBank mRNA AK126166.1 - 4 3003
          BC006441.1 - 4 1924
          BC112115.1 - 4 1053
          BC112117.1 - 4 1053
          BC143685.1 - 4 1041
          Gene Symbol: RRN3P2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003369.2 12 1271
          GenBank mRNA AK225670.1 - 12 1121
          AK298442.1 - 10 1012

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          general transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          transcription initiation from RNA polymerase I promoter
          nucleolus organization
          cytoplasm organization
          cell proliferation
          positive regulation of neuron projection development
          ribosome biogenesis
          positive regulation of transcription, DNA-templated
          homeostasis of number of cells
          negative regulation of intrinsic apoptotic signaling pathway by p53 class mediator
          regulation of DNA-templated transcription, initiation
          RNA polymerase I CORE element sequence-specific DNA binding
          RNA polymerase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0