Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PIH1D1 SIRNA Assays ›
          Gene Symbol:
          PIH1D1
          Gene Name:
          PIH1 domain containing 1
          Gene Aliases:
          NOP17
          Chromosome Location:
          Chr.19: 49446293 - 49452332 on Build GRCh38
          UniGene ID:
          Hs.5245
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s29985

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PIH1D1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_017916.2 NP_060386.1 3 465
          XM_006723254.2 XP_006723317.1 2 91
          GenBank mRNA AK000650.1 - 3 417
          AK294687.1 - 3 391
          AK304476.1 - 3 516
          BC001108.2 - 3 395 2966990
          DC385084.1 - 3 465

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          box C/D snoRNP assembly
          chromatin remodeling
          transcription, DNA-templated
          epithelial cell differentiation
          positive regulation of protein complex assembly
          snoRNA localization
          regulation of histone H3-K4 methylation
          establishment of protein localization to chromatin
          positive regulation of protein serine/threonine kinase activity
          positive regulation of histone H4 acetylation
          negative regulation of histone H3-K9 dimethylation
          negative regulation of histone H3-K9 trimethylation
          positive regulation of transcription of nuclear large rRNA transcript from RNA polymerase I promoter
          positive regulation of glucose mediated signaling pathway
          regulation of TORC2 signaling
          positive regulation of TORC1 signaling
          positive regulation of histone H3-K9 acetylation
          negative regulation of histone H4-K16 acetylation
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
          RNA polymerase I CORE element sequence-specific DNA binding
          RNA polymerase I upstream control element sequence-specific DNA binding
          protein binding
          protein kinase binding
          histone binding
          ATPase binding
          phosphoprotein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-p2dpg:80/100.66.77.81:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0