Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARHGEF10L SIRNA Assays ›
Gene Symbol:
ARHGEF10L
Gene Name:
Rho guanine nucleotide exchange factor 10 like
Gene Aliases:
GrinchGEF
Chromosome Location:
Chr.1: 17520197 - 17697875 on Build GRCh38
UniGene ID:
Hs.443460
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s30314
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARHGEF10L
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001011722.2
NP_001011722.2
17, 18
1966
NM_001319837.1
NP_001306766.1
16, 17
1951
NM_001319838.1
NP_001306767.1
8, 9
1190
NM_001328124.1
NP_001315053.1
11, 12
1415
NM_018125.3
NP_060595.3
19, 20
2199
NR_137287.1
19, 20
2202
NR_137288.1
18, 19
2082
XM_005245923.2
XP_005245980.2
18, 19
2058
XM_005245925.2
XP_005245982.2
18, 19
2046
XM_005245927.2
XP_005245984.2
11, 12
1415
XM_005245929.2
XP_005245986.2
10, 11
1400
XM_006710728.1
XP_006710791.1
18, 19
2240
XM_006710729.1
XP_006710792.1
18, 19
2055
XM_006710731.1
XP_006710794.1
18, 19
2237
XM_011541691.2
XP_011539993.1
17, 18
2055
XM_011541692.2
XP_011539994.1
16, 17
2049
XM_011541693.2
XP_011539995.1
16, 17
2040
XM_011541695.2
XP_011539997.1
17, 18
2064
XM_011541696.2
XP_011539998.1
16, 17
2049
XM_017001617.1
XP_016857106.1
17, 18
2031
XM_017001618.1
XP_016857107.1
12, 13
1333
XM_017001619.1
XP_016857108.1
17, 18
2064
XM_017001620.1
XP_016857109.1
16, 17
2049
XM_017001621.1
XP_016857110.1
18, 19
2182
XM_017001622.1
XP_016857111.1
17, 18
2167
GenBank mRNA
AB046846.1
-
19, 20
2199
AK001383.1
-
5, 6
657
AK092020.1
-
8, 9
1187
AK126877.1
-
10, 11
2305
AK300174.1
-
15, 16
1730
AK300264.1
-
10, 11
1363
AK311076.1
-
18, 19
2009
BC065561.1
-
5, 6
701
BC080596.1
-
8, 9
862
BC117171.1
-
17, 18
1956
40125722
BC143347.1
-
17, 18
1956
9051852
DQ139833.1
-
16, 17
1908
Back To Top
More Information
Panther Classification:
Molecular Function -
G-protein modulator
protein-binding activity modulator
guanyl-nucleotide exchange factor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of Rho protein signal transduction
positive regulation of GTPase activity
Rho guanyl-nucleotide exchange factor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0