Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ALG13 SIRNA Assays ›
Gene Symbol:
ALG13
Gene Name:
ALG13, UDP-N-acetylglucosaminyltransferase subunit
Gene Aliases:
CDG1S, CXorf45, EIEE36, GLT28D1, MDS031, TDRD13, YGL047W
Chromosome Location:
Chr.X: 111680741 - 111764164 on Build GRCh38
UniGene ID:
Hs.443061
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s31655
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
392.00
Your Price
Online offer:
380.65
380.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ALG13
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001039210.4
NP_001034299.3
2
259
NM_001099922.2
NP_001093392.1
2
259
NM_001168385.2
NP_001161857.1
2
259
NM_001257231.1
NP_001244160.1
2
332
NM_001257237.1
NP_001244166.1
2
332
NM_001257239.2
NP_001244168.1
2
247
NM_001257240.2
NP_001244169.1
2
332
NM_001257241.2
NP_001244170.1
2
332
NM_001324290.1
NP_001311219.1
2
259
NM_001324292.1
NP_001311221.1
2
259
NM_001324293.1
NP_001311222.1
2
332
NM_018466.5
NP_060936.1
2
259
XM_006724693.2
XP_006724756.1
2
632
XM_011531028.2
XP_011529330.1
2
633
XM_011531030.1
XP_011529332.1
2
631
XM_011531031.1
XP_011529333.1
2
332
XM_011531033.2
XP_011529335.1
2
178
XM_017029846.1
XP_016885335.1
2
634
XM_017029847.1
XP_016885336.1
2
633
XM_017029848.1
XP_016885337.1
2
631
XM_017029849.1
XP_016885338.1
2
632
XM_017029850.1
XP_016885339.1
2
631
XM_017029851.1
XP_016885340.1
2
634
GenBank mRNA
AF220051.1
-
2
190
AK302729.1
-
2
200
AK302890.1
-
2
235
AK304712.1
-
2
149
AK311608.1
-
2
198
AK312229.1
-
2
198
AK316522.1
-
2
276
BC005336.1
-
2
192
3996922
BE747995.1
-
2
188
BF239206.1
-
2
286
BF699152.1
-
2
260
BG621042.1
-
1
42
BM759764.1
-
2
241
DA830058.1
-
2
323
DB292609.1
-
2
332
DN992019.1
-
2
327
Back To Top
More Information
Panther Classification:
Molecular Function -
cysteine protease
transferase
Gene Ontology Categories:
Function(s)
Process(es)
dolichol-linked oligosaccharide biosynthetic process
proteolysis
N-acetylglucosaminyldiphosphodolichol N-acetylglucosaminyltransferase activity
thiol-dependent ubiquitinyl hydrolase activity
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-4p4qf:80/100.66.72.111:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0