Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ALG13 SIRNA Assays ›
          Gene Symbol:
          ALG13
          Gene Name:
          ALG13, UDP-N-acetylglucosaminyltransferase subunit
          Gene Aliases:
          CDG1S, CXorf45, EIEE36, GLT28D1, MDS031, TDRD13, YGL047W
          Chromosome Location:
          Chr.X: 111680741 - 111764164 on Build GRCh38
          UniGene ID:
          Hs.443061
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s31656

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ALG13
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001099922.2 NP_001093392.1 2 298
          NM_001168385.2 NP_001161857.1 2 298
          NM_001257230.1 NP_001244159.1 2 194
          NM_001257234.1 NP_001244163.1 2 267
          NM_001257235.2 NP_001244164.1 2 267
          NM_001257237.1 NP_001244166.1 2 371
          NM_001257239.2 NP_001244168.1 2 286
          NM_001257240.2 NP_001244169.1 2 371
          NM_001324290.1 NP_001311219.1 2 298
          NM_001324291.1 NP_001311220.1 2 194
          NM_001324292.1 NP_001311221.1 2 298
          NM_001324293.1 NP_001311222.1 2 371
          NM_001324294.1 NP_001311223.1 2 182
          NM_018466.5 NP_060936.1 2 298
          NR_033125.2 2 194
          XM_006724693.2 XP_006724756.1 2 671
          XM_006724697.3 XP_006724760.2 2 571
          XM_006724698.3 XP_006724761.2 2 696
          XM_011531028.2 XP_011529330.1 2 672
          XM_011531030.1 XP_011529332.1 2 670
          XM_011531031.1 XP_011529333.1 2 371
          XM_011531032.1 XP_011529334.1 2 177
          XM_011531033.2 XP_011529335.1 2 217
          XM_017029846.1 XP_016885335.1 2 673
          XM_017029847.1 XP_016885336.1 2 672
          XM_017029848.1 XP_016885337.1 2 670
          XM_017029849.1 XP_016885338.1 2 671
          XM_017029850.1 XP_016885339.1 2 670
          XM_017029851.1 XP_016885340.1 2 673
          GenBank mRNA AF220051.1 - 2 229
          AK300394.1 - 2 136
          AK302729.1 - 2 239
          AK304712.1 - 2 188
          AK312229.1 - 2 237
          AK316306.1 - 2 240
          AK316522.1 - 2 315
          AL524204.3 - 2 117
          BC005336.1 - 2 231 3996922
          BE747995.1 - 2 227
          BF240663.1 - 2 85
          BF699152.1 - 2 299
          BG621042.1 - 1 81
          BM759764.1 - 2 280
          CB268871.1 - 2 245
          DA830058.1 - 2 362
          DB292609.1 - 2 371
          DC400909.1 - 2 182
          DN992019.1 - 2 366

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cysteine protease transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          dolichol-linked oligosaccharide biosynthetic process
          proteolysis
          N-acetylglucosaminyldiphosphodolichol N-acetylglucosaminyltransferase activity
          thiol-dependent ubiquitinyl hydrolase activity
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0