Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: OXCT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022120.1 | NP_071403.1 | 1 | 1520 | ||
GenBank mRNA | AB050193.2 | - | 1 | 1446 | ||
AK024440.1 | - | 1 | 625 | |||
AK093089.1 | - | 1 | 1494 | |||
AK314586.1 | - | 1 | 1427 | |||
AY013700.1 | - | 1 | 1520 | |||
BC030015.1 | - | 1 | 1439 |
Gene Symbol: OXCT2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132962.1 | 1 | 1498 | |||
GenBank mRNA | BC072388.1 | - | 1 | 1498 | 5588771 |