Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SEH1L SIRNA Assays ›
          Gene Symbol:
          SEH1L
          Gene Name:
          SEH1 like nucleoporin
          Gene Aliases:
          SEC13L, SEH1A, SEH1B, Seh1
          Chromosome Location:
          Chr.18: 12947984 - 12987665 on Build GRCh38
          UniGene ID:
          Hs.301048
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s37880

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SEH1L
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001013437.1 NP_001013455.1 7 960
          NM_031216.3 NP_112493.2 7 960
          XM_005258152.4 XP_005258209.1 6 803
          XM_011525742.2 XP_011524044.1 8 1016
          XM_011525743.2 XP_011524045.1 7 863
          XM_011525744.1 XP_011524046.1 8 1016
          XM_011525745.2 XP_011524047.1 7 784
          XM_011525746.2 XP_011524048.1 7 1066
          XM_017026025.1 XP_016881514.1 6 809
          XM_017026026.1 XP_016881515.1 6 880
          XM_017026027.1 XP_016881516.1 7 1130
          GenBank mRNA AF136976.1 - 7 928
          AF255625.1 - 7 864
          AF431970.1 - 7 864
          AF514996.1 - 7 824
          AK056940.1 - 7 923
          AK226075.1 - 4 3350
          AK291226.1 - 7 923
          AK314403.1 - 7 934
          BC012430.1 - 7 914 3865994
          BX538037.1 - 2 2669

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          cytokine production involved in inflammatory response
          mRNA export from nucleus
          tRNA export from nucleus
          nuclear pore organization
          sister chromatid cohesion
          mitotic nuclear envelope disassembly
          mitotic metaphase plate congression
          regulation of glucose transport
          protein transport
          viral process
          protein sumoylation
          viral transcription
          gene silencing by RNA
          positive regulation of TOR signaling
          cellular response to amino acid starvation
          cellular protein complex localization
          defense response to Gram-positive bacterium
          cell division
          attachment of mitotic spindle microtubules to kinetochore
          intracellular transport of virus
          regulation of cellular response to heat
          positive regulation of TORC1 signaling
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0