Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TCHHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008536.1 | NP_001008536.1 | 3 | 2459 | ||
GenBank mRNA | AY456639.1 | - | 3 | 2459 | ||
BC137493.1 | - | 3 | 2449 | 9021493 |