Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PLK1 SIRNA Assays ›
          Gene Symbol:
          PLK1
          Gene Name:
          polo like kinase 1
          Gene Aliases:
          PLK, STPK13
          Chromosome Location:
          Chr.16: 23678772 - 23690367 on Build GRCh38
          UniGene ID:
          Hs.592049
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s449
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 7,898.00
          Your Price
          Online offer:
          7898.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PLK1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005030.5 NP_005021.2 3 762
          GenBank mRNA AB084459.1 - 3 605
          AB209179.1 - 2 2621
          AK303263.1 - 4 378
          AK313227.1 - 3 686
          BC002369.2 - 3 654 2822226
          BC003002.1 - 3 632 2823623
          BC014846.1 - 3 607 3854860
          L19559.1 - 3 664
          U01038.1 - 3 664
          X73458.1 - 3 678
          X75932.1 - 3 664

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00153444_m1

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic sister chromatid segregation
          G2/M transition of mitotic cell cycle
          negative regulation of transcription from RNA polymerase II promoter
          mitotic cytokinesis
          cytokinesis
          microtubule bundle formation
          protein phosphorylation
          sister chromatid cohesion
          mitotic nuclear division
          mitotic nuclear envelope disassembly
          mitotic spindle assembly checkpoint
          regulation of mitotic cell cycle
          cell proliferation
          positive regulation of peptidyl-threonine phosphorylation
          female meiosis chromosome segregation
          protein ubiquitination
          peptidyl-serine phosphorylation
          regulation of mitotic metaphase/anaphase transition
          anaphase-promoting complex-dependent catabolic process
          G2 DNA damage checkpoint
          protein destabilization
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          negative regulation of apoptotic process
          regulation of protein binding
          homologous chromosome segregation
          establishment of protein localization
          negative regulation of cyclin-dependent protein serine/threonine kinase activity
          positive regulation of proteolysis
          centrosome organization
          positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
          regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
          positive regulation of ubiquitin-protein transferase activity
          regulation of cell cycle
          synaptonemal complex disassembly
          protein localization to chromatin
          positive regulation of protein localization to nucleus
          regulation of mitotic spindle assembly
          regulation of cell cycle G2/M phase transition
          positive regulation of ubiquitin protein ligase activity
          protein kinase activity
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          microtubule binding
          anaphase-promoting complex binding
          kinase activity
          protein kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0