Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DMTN SIRNA Assays ›
Gene Symbol:
DMTN
Gene Name:
dematin actin binding protein
Gene Aliases:
DMT, EPB49
Chromosome Location:
Chr.8: 22048955 - 22082527 on Build GRCh38
UniGene ID:
Hs.106124
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s4713
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DMTN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001114135.4
NP_001107607.1
10
1056
NM_001114136.2
NP_001107608.1
10
1012
NM_001114137.3
NP_001107609.1
10
1186
NM_001114138.2
NP_001107610.1
10
1173
NM_001114139.3
NP_001107611.1
9
1070
NM_001302816.2
NP_001289745.1
10
1306
NM_001302817.2
NP_001289746.1
8
1025
NM_001323378.1
NP_001310307.1
10
1145
NM_001323380.1
NP_001310309.1
10
1052
NM_001323381.1
NP_001310310.1
9
1161
NM_001323382.1
NP_001310311.1
10
1091
NM_001323383.1
NP_001310312.1
10
1012
NM_001323384.1
NP_001310313.1
10
1145
NM_001323385.1
NP_001310314.1
9
1161
NM_001323387.1
NP_001310316.1
9
1110
NM_001323388.1
NP_001310317.1
8
892
NM_001323389.1
NP_001310318.1
9
1100
NM_001323390.1
NP_001310319.1
9
967
NM_001323391.1
NP_001310320.1
9
1070
NM_001323392.1
NP_001310321.1
9
1231
NM_001323393.1
NP_001310322.1
9
937
NM_001323394.1
NP_001310323.1
9
1098
NM_001323395.1
NP_001310324.1
9
977
NM_001323396.1
NP_001310325.1
9
981
NM_001323397.1
NP_001310326.1
9
1149
NM_001323398.1
NP_001310327.1
8
1025
NM_001323399.1
NP_001310328.1
8
892
NM_001323400.1
NP_001310329.1
7
1041
NM_001323401.1
NP_001310330.1
8
1065
NM_001978.4
NP_001969.2
10
1306
XM_005273432.1
XP_005273489.1
10
1038
XM_005273433.1
XP_005273490.1
10
970
XM_005273442.1
XP_005273499.1
9
822
XM_011544435.1
XP_011542737.1
9
1231
XM_017013189.1
XP_016868678.1
10
979
XM_017013190.1
XP_016868679.1
9
1231
XM_017013191.1
XP_016868680.1
9
985
XM_017013192.1
XP_016868681.1
9
866
XM_017013193.1
XP_016868682.1
8
998
XM_017013194.1
XP_016868683.1
8
865
XM_017013195.1
XP_016868684.1
9
1231
XM_017013196.1
XP_016868685.1
9
834
XM_017013197.1
XP_016868686.1
9
1065
XM_017013198.1
XP_016868687.1
9
907
GenBank mRNA
AB209718.1
-
6
532
AK055842.1
-
10
970
AK091581.1
-
9
985
AK289650.1
-
10
938
AK295452.1
-
8
911
BC006318.2
-
10
1055
4125804
BC017445.1
-
10
1055
3162732
BC052805.1
-
10
1096
6269658
BT007396.1
-
9
783
L19713.1
-
10
1233
U28389.1
-
10
1233
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
protein complex assembly
cytoskeleton organization
regulation of cell shape
regulation of lamellipodium assembly
positive regulation of fibroblast migration
negative regulation of peptidyl-threonine phosphorylation
negative regulation of cell-substrate adhesion
actin cytoskeleton organization
positive regulation of blood coagulation
regulation of actin cytoskeleton organization
negative regulation of peptidyl-serine phosphorylation
calcium-mediated signaling using intracellular calcium source
calcium-mediated signaling using extracellular calcium source
erythrocyte development
negative regulation of peptidyl-tyrosine phosphorylation
actin filament bundle assembly
regulation of filopodium assembly
actin filament capping
negative regulation of focal adhesion assembly
transmembrane transport
protein secretion by platelet
cellular response to calcium ion
cellular response to cAMP
positive regulation of wound healing
negative regulation of protein targeting to membrane
actin filament reorganization
negative regulation of substrate adhesion-dependent cell spreading
positive regulation of substrate adhesion-dependent cell spreading
positive regulation of platelet aggregation
positive regulation of integrin-mediated signaling pathway
actin binding
receptor binding
protein binding
spectrin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-jgtjs:80/10.239.162.121:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0