Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CNEP1R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281789.1 | NP_001268718.1 | 4 | 331 | ||
NM_153261.5 | NP_694993.2 | 5 | 382 | |||
NR_104042.1 | 4 | 308 | ||||
NR_104043.1 | 3 | 257 | ||||
GenBank mRNA | AK095420.1 | - | 4 | 304 | ||
AK301610.1 | - | 5 | 347 | |||
AK307132.1 | - | 4 | 378 | |||
AK307309.1 | - | 3 | 199 | |||
BC022550.1 | - | 3 | 208 | |||
BC036683.2 | - | 5 | 311 | 5266084 | ||
BP259769.1 | - | 4 | 280 | |||
CR933600.1 | - | 5 | 327 | |||
FJ516381.1 | - | 4 | 259 | |||
HY011229.1 | - | 4 | 332 |