Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LRRC37A3 SIRNA Assays ›
Gene Symbol:
LRRC37A3
Gene Name:
leucine rich repeat containing 37 member A3
Gene Aliases:
LRRC37, LRRC37A
Chromosome Location:
Chr.17: 64854147 - 64966228 on Build GRCh38
UniGene ID:
Hs.551962
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s51586
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LRRC37A3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001303255.1
NP_001290184.1
8
1307
NM_199340.3
NP_955372.2
12
4564
XM_011524776.1
XP_011523078.1
9
2003
XM_011524777.1
XP_011523079.1
7
1850
XM_011524778.2
XP_011523080.2
10
1528
XM_017024593.1
XP_016880082.1
10
4393
XM_017024594.1
XP_016880083.1
9
4321
XM_017024595.1
XP_016880084.1
9
4321
XM_017024596.1
XP_016880085.1
8
4240
XM_017024597.1
XP_016880086.1
8
4240
XM_017024598.1
XP_016880087.1
7
4168
XM_017024599.1
XP_016880088.1
11
3765
XM_017024601.1
XP_016880090.1
9
1705
XM_017024602.1
XP_016880091.1
6
1480
XM_017024603.1
XP_016880092.1
9
2003
XM_017024604.1
XP_016880093.1
8
1931
XM_017024605.1
XP_016880094.1
7
1850
XM_017024606.1
XP_016880095.1
6
1778
XM_017024607.1
XP_016880096.1
9
1308
XM_017024608.1
XP_016880097.1
9
2325
GenBank mRNA
AB011135.2
-
11
4429
AK091625.1
-
2
1412
AK294371.1
-
8
1295
AK299714.1
-
5
1061
AK308554.1
-
5
1040
AK315945.1
-
9
2036
BC043145.1
-
9
3906
5296147
BC113643.1
-
2
926
BC117274.1
-
2
926
Back To Top
More Information
Panther Classification:
Molecular Function -
transmembrane signal receptor
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0