Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CCR5 SIRNA Assays ›
          Gene Symbol:
          CCR5
          Gene Name:
          C-C motif chemokine receptor 5 (gene/pseudogene)
          Gene Aliases:
          CC-CKR-5, CCCKR5, CCR-5, CD195, CKR-5, CKR5, CMKBR5, IDDM22
          Chromosome Location:
          Chr.3: 46370142 - 46376206 on Build GRCh38
          UniGene ID:
          Hs.450802
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s531495

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CCR5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000579.3 NP_000570.1 3 1120
          NM_001100168.1 NP_001093638.1 3 885
          GenBank mRNA AF011500.1 - 1 767
          AF011501.1 - 1 767
          AF011502.1 - 1 767
          AF011503.1 - 1 767
          AF011504.1 - 1 766
          AF011505.1 - 1 767
          AF011506.1 - 1 767
          AF011507.1 - 1 767
          AF011508.1 - 1 767
          AF011510.1 - 1 767
          AF011511.1 - 1 767
          AF011512.1 - 1 767
          AF011513.1 - 1 767
          AF011514.1 - 1 767
          AF011515.1 - 1 767
          AF011516.1 - 1 767
          AF011517.1 - 1 767
          AF011518.1 - 1 767
          AF011519.1 - 1 767
          AF011520.1 - 1 767
          AF011521.1 - 1 767
          AF011522.1 - 1 767
          AF011523.1 - 1 767
          AF011524.1 - 1 767
          AF011525.1 - 1 767
          AF011526.1 - 1 767
          AF011527.1 - 1 767
          AF011528.1 - 1 767
          AF011529.1 - 1 767
          AF011530.1 - 1 767
          AF011531.1 - 1 767
          AF011532.1 - 1 767
          AF011533.1 - 1 767
          AF011534.1 - 1 767
          AF011535.1 - 1 767
          AF011536.1 - 1 767
          AF011537.1 - 1 767
          AK292132.1 - 2 817
          AK314882.1 - 3 877
          BC038398.1 - 2 806 5186388
          U54994.1 - 2 817
          U57840.1 - 2 789

          Back To Top

          More Information


          Set Membership:

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          dendritic cell chemotaxis
          calcium ion transport
          chemotaxis
          inflammatory response
          immune response
          cellular defense response
          cell surface receptor signaling pathway
          G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          cell-cell signaling
          release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
          fusion of virus membrane with host plasma membrane
          calcium-mediated signaling
          signaling
          entry into host cell
          chemokine-mediated signaling pathway
          response to cholesterol
          cellular response to lipopolysaccharide
          virus receptor activity
          actin binding
          phosphatidylinositol phospholipase C activity
          chemokine receptor activity
          protein binding
          coreceptor activity
          C-C chemokine receptor activity
          C-C chemokine binding
          chemokine (C-C motif) ligand 5 binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-dhnrf:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0