Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PPOX SIRNA Assays ›
          Gene Symbol:
          PPOX
          Gene Name:
          protoporphyrinogen oxidase
          Gene Aliases:
          PPO, V290M, VP
          Chromosome Location:
          Chr.1: 161165728 - 161178277 on Build GRCh38
          UniGene ID:
          Hs.517373
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s533799

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 392.00
          Your Price
          Online offer: 380.65
          380.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PPOX
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000309.3 NP_000300.1 7 932
          NM_001122764.1 NP_001116236.1 7 993
          XM_005245291.3 XP_005245348.2 7 971
          XM_005245295.3 XP_005245352.2 6 819
          XM_006711402.2 XP_006711465.2 7 938
          XM_006711403.2 XP_006711466.2 7 927
          XM_006711404.3 XP_006711467.2 7 875
          XM_006711406.2 XP_006711469.2 7 848
          XM_011509663.2 XP_011507965.1 7 1058
          XM_011509664.1 XP_011507966.1 7 829
          XM_011509665.2 XP_011507967.1 7 1056
          XM_011509666.2 XP_011507968.1 7 1063
          XM_011509667.2 XP_011507969.1 7 999
          XM_011509668.2 XP_011507970.1 7 799
          XM_011509669.1 XP_011507971.1 7 812
          XM_011509670.2 XP_011507972.1 6 947
          XM_011509671.1 XP_011507973.1 7 829
          XM_011509672.2 XP_011507974.1 6 938
          XM_011509673.2 XP_011507975.1 5 795
          XM_011509674.2 XP_011507976.1 7 1064
          XM_011509675.2 XP_011507977.1 6 880
          XM_011509676.1 XP_011507978.1 6 1143
          XM_011509677.1 XP_011507979.1 6 1076
          XM_011509678.1 XP_011507980.1 7 1036
          XM_011509679.1 XP_011507981.1 7 844
          XM_011509681.1 XP_011507983.1 6 761
          XM_011509682.1 XP_011507984.1 6 771
          XM_017001559.1 XP_016857048.1 7 1055
          XM_017001560.1 XP_016857049.1 7 971
          XM_017001561.1 XP_016857050.1 6 1464
          XM_017001562.1 XP_016857051.1 7 917
          XM_017001563.1 XP_016857052.1 7 1015
          XM_017001564.1 XP_016857053.1 3 533
          XM_017001565.1 XP_016857054.1 7 917
          XM_017001566.1 XP_016857055.1 7 1026
          XM_017001567.1 XP_016857056.1 6 819
          XM_017001568.1 XP_016857057.1 7 1007
          XM_017001569.1 XP_016857058.1 6 761
          XM_017001570.1 XP_016857059.1 4 562
          GenBank mRNA AK094855.1 - 6 765
          AK302298.1 - 6 803
          BC002357.2 - 7 913 2821983
          D38537.1 - 7 980
          U26446.1 - 7 894

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          oxidase

          Gene Ontology Categories:

          Function(s) Process(es)

          porphyrin-containing compound biosynthetic process
          protoporphyrinogen IX biosynthetic process
          heme biosynthetic process
          response to drug
          oxidation-reduction process
          oxygen-dependent protoporphyrinogen oxidase activity
          flavin adenine dinucleotide binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0