Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CDK5RAP2 SIRNA Assays ›
          Gene Symbol:
          CDK5RAP2
          Gene Name:
          CDK5 regulatory subunit associated protein 2
          Gene Aliases:
          C48, Cep215, MCPH3
          Chromosome Location:
          Chr.9: 120388869 - 120580170 on Build GRCh38
          UniGene ID:
          Hs.269560
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s534188

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CDK5RAP2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001011649.2 NP_001011649.1 24 3505
          NM_001272039.1 NP_001258968.1 21 2815
          NM_018249.5 NP_060719.4 24 3505
          NR_073554.1 24 3582
          NR_073555.1 24 3505
          NR_073556.1 25 3712
          NR_073557.1 24 3585
          NR_073558.1 24 3582
          XM_006717182.1 XP_006717245.1 23 3398
          XM_006717185.1 XP_006717248.1 21 2807
          XM_011518860.1 XP_011517162.1 24 3491
          XM_017014921.1 XP_016870410.1 23 3395
          XM_017014922.1 XP_016870411.1 18 2723
          XM_017014923.1 XP_016870412.1 21 2807
          XM_017014924.1 XP_016870413.1 15 2479
          GenBank mRNA AB046853.2 - 24 3474
          AF448860.1 - 24 3455
          AK001729.1 - 2 343
          AK027636.1 - 3 469
          AK122913.1 - 24 3494
          AL133161.1 - 8 1495
          AL833481.1 - 24 3493
          BC136275.1 - 24 3337 9020262
          BC143762.1 - 24 3340 9052276
          BC146782.1 - 24 3474 9007237
          BX537421.1 - 24 3490
          BX537708.1 - 21 2804
          BX537759.1 - 24 3493
          BX640896.1 - 17 2621
          CR936622.1 - 17 2635
          CR936717.1 - 17 2635
          KP294527.1 - 24 3505

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          establishment of mitotic spindle orientation
          microtubule cytoskeleton organization
          microtubule bundle formation
          chromosome segregation
          centriole replication
          brain development
          neurogenesis
          microtubule organizing center organization
          regulation of neuron differentiation
          negative regulation of neuron differentiation
          positive regulation of transcription, DNA-templated
          negative regulation of centriole replication
          centrosome organization
          regulation of spindle checkpoint
          protein binding
          calmodulin binding
          microtubule binding
          tubulin binding
          protein kinase binding
          protein complex binding
          transcription regulatory region DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-mfr4m:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0