Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ADH1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000669.4 | NP_000660.1 | 9 | 1619 | ||
NR_133005.1 | 9 | 1575 | ||||
XM_011531588.2 | XP_011529890.1 | 8 | 1409 | |||
XM_011531589.1 | XP_011529891.1 | 10 | 1739 | |||
GenBank mRNA | BC062476.1 | - | 9 | 1315 | 30408761 | |
CD014078.1 | - | 9 | 1276 | |||
M12272.1 | - | 9 | 1329 | |||
X04299.1 | - | 9 | 1321 | |||
X04350.1 | - | 7 | 1091 |