Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SLC4A11 SIRNA Assays ›
          Gene Symbol:
          SLC4A11
          Gene Name:
          solute carrier family 4 member 11
          Gene Aliases:
          BTR1, CDPD1, CHED, CHED2, NABC1, dJ794I6.2
          Chromosome Location:
          Chr.20: 3227417 - 3241483 on Build GRCh38
          UniGene ID:
          Hs.105607
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s536678

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SLC4A11
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001174089.1 NP_001167560.1 9, 10 1155
          NM_001174090.1 NP_001167561.1 9, 10 1305
          NM_032034.3 NP_114423.1 8, 9 1147
          NR_135000.1 9, 10 1219
          XM_005260856.4 XP_005260913.1 9, 10 1643
          XM_005260857.1 XP_005260914.1 9, 10 1174
          XM_011529383.2 XP_011527685.1 9, 10 2750
          XM_011529384.1 XP_011527686.1 9, 10 1392
          XM_011529385.1 XP_011527687.1 9, 10 1219
          XM_017028093.1 XP_016883582.1 9, 10 1643
          XM_017028094.1 XP_016883583.1 9, 10 1027
          XM_017028095.1 XP_016883584.1 9, 10 1498
          XM_017028096.1 XP_016883585.1 10, 11 1404
          XM_017028097.1 XP_016883586.1 9, 10 1643
          GenBank mRNA AF336127.1 - 8, 9 1147
          AK075303.1 - 3, 4 422
          AK091157.1 - 9, 10 1219
          AK296508.1 - 9, 10 1305
          AK296760.1 - 9, 10 1104
          BC110540.1 - 8, 9 1120 40035522
          BC110541.1 - 8, 9 1120

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          sodium ion transport
          bicarbonate transport
          proton transport
          cellular cation homeostasis
          borate transmembrane transport
          sodium ion transmembrane transport
          fluid transport
          borate transport
          regulation of intracellular pH
          sodium channel activity
          inorganic anion exchanger activity
          bicarbonate transmembrane transporter activity
          hydrogen ion channel activity
          symporter activity
          anion:anion antiporter activity
          borate transmembrane transporter activity
          protein dimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0