Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TARP SIRNA Assays ›
          Gene Symbol:
          TARP TRGC2 TRGV9
          Gene Name:
          TCR gamma alternate reading frame protein
          T cell receptor gamma constant 2
          T cell receptor gamma variable 9
          Gene Aliases:
          CD3G, TCRG, TCRGC1, TCRGC2, TCRGV
          TCRGC2, TRGC2(2X), TRGC2(3X)
          TCRGV9, TRGC1, V2
          Chromosome Location:
          Chr.7: 38239577 - 38276741 on Build GRCh38
          Chromosome Location:
          Chr.7: 38240024 - 38249572 on Build GRCh38
          Chromosome Location:
          Chr.7: 38317017 - 38317492 on Build GRCh38
          UniGene ID:
          Hs.534032
          Hs.731375
          Hs.534032
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID s54579

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TARP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001003799.1 NP_001003799.1 3 403
          NM_001003806.1 NP_001003806.1 3 403
          XM_011515407.2 XP_011513709.1 4 623
          XM_017012226.1 XP_016867715.1 2 180
          XM_017012227.1 XP_016867716.1 3 613
          XM_017012228.1 XP_016867717.1 2 54
          XM_017012229.1 XP_016867718.1 3 825
          XM_017012230.1 XP_016867719.1 3 429
          GenBank mRNA AF151103.1 - 3 403
          BC030554.1 - 5 1087
          BC105589.1 - 4 621 4272944
          BF674593.1 - 3 393
          KC170727.1 - 6 760
          M16804.1 - 6 778
          M17324.1 - 3 390
          M17325.1 - 2 322
          M27331.1 - 3 391
          M27332.1 - 3 391
          M27334.1 - 2 343
          M30894.1 - 978
          M33028.1 - 1 12
          X72500.1 - 5 769
          Y00790.1 - 6 829
          Gene Symbol: TRGC2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA M13231.1 - 6 800
          M17323.1 - 438
          Gene Symbol: TRGV9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC072387.1 - 4 720
          BC072396.1 - 6 1146
          M16768.1 - 5 906

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0