Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TAF9B SIRNA Assays ›
          Gene Symbol:
          TAF9B
          Gene Name:
          TATA-box binding protein associated factor 9b
          Gene Aliases:
          DN-7, DN7, TAF9L, TAFII31L, TFIID-31
          Chromosome Location:
          Chr.X: 78129748 - 78139682 on Build GRCh38
          UniGene ID:
          Hs.592248
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s58379

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TAF9B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_015975.4 NP_057059.2 3 261
          GenBank mRNA AF220509.1 - 3 212
          AK312579.1 - 3 220
          BC009566.1 - 3 212 3996687
          BC010350.1 - 3 226 4276335
          BC071649.1 - 3 198 5246715
          BC146952.1 - 3 242 9056880
          BC146960.1 - 3 242 9056888
          BI544376.1 - 3 289

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          transcription initiation from RNA polymerase II promoter
          transcription elongation from RNA polymerase II promoter
          positive regulation of cell growth
          negative regulation of apoptotic process
          protein stabilization
          regulation of signal transduction by p53 class mediator
          negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          transcription corepressor activity
          protein binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0