Search
Search

Chrng| Gene Symbol: Chrng | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009604.3 | NP_033734.3 | 4 | 305 | ||
| GenBank mRNA | AK047974.1 | - | 4 | 306 | ||
| JF520813.1 | - | 4 | 306 | |||
| JN164665.1 | - | 473 | ||||
| JN164666.1 | - | 3 | 364 | |||
| M30514.1 | - | 4 | 346 | |||
| X03818.1 | - | 4 | 302 | |||
| X03819.1 | - | 3 | 237 | |||