Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CORO1A SIRNA Assays ›
          Gene Symbol:
          Coro1a
          Gene Name:
          coronin, actin binding protein 1A
          Gene Aliases:
          Clabp, Lmb3, TACO, p57
          Chromosome Location:
          Chr.7: 126699774 - 126704816 on Build GRCm38
          UniGene ID:
          Mm.290482
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s64010

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Coro1a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001301374.1 NP_001288303.1 7 1064
          NM_009898.3 NP_034028.1 7 977
          GenBank mRNA AF047388.1 - 6 811
          AF143955.1 - 7 948
          AF495468.1 - 7 915
          AK040885.1 - 7 921
          AK075799.1 - 7 912
          AK146290.1 - 7 915
          AK150962.1 - 7 919
          AK151755.1 - 7 919
          AK155527.1 - 7 920
          AK155860.1 - 7 915
          AK165296.1 - 7 919
          AK172447.1 - 7 915
          BC002136.1 - 7 905 3484185
          BC053398.1 - 7 895 30076939
          U89399.1 - 3 347

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-motor actin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          phagolysosome assembly
          calcium ion transport
          phagocytosis
          actin filament organization
          regulation of actin polymerization or depolymerization
          regulation of cell shape
          cell migration
          actin cytoskeleton organization
          positive regulation of cell migration
          leukocyte chemotaxis
          regulation of actin filament polymerization
          negative regulation of vesicle fusion
          cell-substrate adhesion
          uropod organization
          regulation of actin cytoskeleton organization
          response to cytokine
          nerve growth factor signaling pathway
          positive regulation of T cell proliferation
          T cell homeostasis
          natural killer cell degranulation
          negative regulation of neuron apoptotic process
          homeostasis of number of cells within a tissue
          positive regulation of T cell activation
          positive chemotaxis
          negative regulation of actin nucleation
          regulation of release of sequestered calcium ion into cytosol
          early endosome to recycling endosome transport
          cellular response to interleukin-4
          actin binding
          actin monomer binding
          protein binding
          protein C-terminus binding
          cytoskeletal protein binding
          myosin heavy chain binding
          identical protein binding
          protein homodimerization activity
          phosphatidylinositol 3-kinase binding
          poly(A) RNA binding
          actin filament binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-76vs2:80/100.66.76.146:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0