Search
Search

Eps15| Gene Symbol: Eps15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159964.1 | NP_001153436.1 | 11 | 1408 | ||
| NM_007943.3 | NP_031969.1 | 23 | 2357 | |||
| GenBank mRNA | AK083176.1 | - | 11 | 1392 | ||
| AY190610.1 | - | 23 | 2330 | |||
| BC048783.1 | - | 23 | 2331 | 6466448 | ||
| L21768.1 | - | 23 | 2330 | |||