Search
Search

Kcnd2| Gene Symbol: Kcnd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019697.3 | NP_062671.1 | 1 | 1217 | ||
| GenBank mRNA | AB045326.1 | - | 1823 | |||
| AB093280.1 | - | 1 | 1698 | |||
| AF107780.1 | - | 1 | 1027 | |||
| AK032268.1 | - | 2008 | ||||
| BC079667.1 | - | 1 | 1217 | 30356567 | ||