Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rad17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044371.2 | NP_001037836.1 | 6 | 817 | ||
NM_001283011.1 | NP_001269940.1 | 7 | 1022 | |||
NM_011233.3 | NP_035363.2 | 6 | 945 | |||
GenBank mRNA | AF085737.1 | - | 6 | 868 | ||
AJ011923.1 | - | 5 | 718 | |||
AK134277.1 | - | 6 | 772 | |||
AK162016.1 | - | 6 | 804 | |||
AK167358.1 | - | 7 | 965 | |||
BC023939.1 | - | 6 | 822 |