Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163310.1 | NP_001156782.1 | 5 | 468 | ||
NM_009069.4 | NP_033095.1 | 6 | 613 | |||
GenBank mRNA | AK005357.1 | - | 6 | 574 | ||
AK014126.1 | - | 6 | 604 | |||
AK018785.1 | - | 5 | 772 | |||
AK164196.1 | - | 6 | 604 | |||
AK168254.1 | - | 6 | 683 | |||
BC012694.1 | - | 6 | 573 | 4023732 | ||
CX242151.1 | - | 5 | 416 | |||
U71205.1 | - | 6 | 550 |