Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TGFBR3 SIRNA Assays ›
Gene Symbol:
Tgfbr3
Gene Name:
transforming growth factor, beta receptor III
Gene Aliases:
1110036H20Rik, AU015626, AW215636, TBRIII
Chromosome Location:
Chr.5: 107106570 - 107289595 on Build GRCm38
UniGene ID:
Mm.200775
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
s75065
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Tgfbr3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_011578.3
NP_035708.2
8
1364
GenBank mRNA
AF039601.1
-
8
1164
AK133234.1
-
6
874
BC070428.1
-
8
1289
6410554
Back To Top
More Information
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
blood vessel development
vasculogenesis
osteoblast differentiation
in utero embryonic development
blastocyst development
epithelial to mesenchymal transition
liver development
blood vessel remodeling
epicardial cell to mesenchymal cell transition
protein complex assembly
protein phosphorylation
immune response
transforming growth factor beta receptor signaling pathway
transforming growth factor beta receptor complex assembly
positive regulation of cell proliferation
embryo development
negative regulation of epithelial cell migration
negative regulation of epithelial to mesenchymal transition
cell growth
BMP signaling pathway
positive regulation of transforming growth factor beta receptor signaling pathway
negative regulation of transforming growth factor beta receptor signaling pathway
positive regulation of BMP signaling pathway
response to follicle-stimulating hormone
response to prostaglandin E
response to luteinizing hormone
intracellular signal transduction
regulation of protein binding
negative regulation of epithelial cell proliferation
regulation of blood vessel size
positive regulation of NF-kappaB transcription factor activity
negative regulation of cellular component movement
ventricular cardiac muscle tissue morphogenesis
palate development
cardiac muscle cell proliferation
definitive hemopoiesis
cardiac epithelial to mesenchymal transition
definitive erythrocyte differentiation
heart trabecula formation
pathway-restricted SMAD protein phosphorylation
coronary vasculature development
coronary vasculature morphogenesis
vasculogenesis involved in coronary vascular morphogenesis
visceral serous pericardium development
negative regulation of apoptotic process involved in morphogenesis
transforming growth factor beta-activated receptor activity
type II transforming growth factor beta receptor binding
transforming growth factor beta receptor binding
protein binding
glycosaminoglycan binding
heparin binding
coreceptor activity
fibroblast growth factor binding
PDZ domain binding
SMAD binding
activin binding
transforming growth factor beta binding
transforming growth factor beta receptor activity, type III
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-sj4nz:80/100.66.77.235:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0