Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ufd1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011672.4 | NP_035802.3 | 3, 4 | 300 | ||
GenBank mRNA | AK010461.1 | - | 3, 4 | 269 | ||
AK012543.1 | - | 1, 2 | 247 | |||
AK088823.1 | - | 3, 4 | 269 | |||
AK147138.1 | - | 3, 4 | 283 | |||
AK169062.1 | - | 3, 4 | 282 | |||
BC006630.1 | - | 3, 4 | 257 | 3708320 | ||
CJ053815.1 | - | 3, 4 | 269 | |||
CJ085208.1 | - | 3, 4 | 300 | |||
U64445.1 | - | 3, 4 | 285 |