Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pxmp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021534.3 | NP_067509.1 | 2, 3 | 222 | ||
GenBank mRNA | AJ271476.1 | - | 2, 3 | 211 | ||
AK003127.1 | - | 2, 3 | 237 | |||
AK013793.1 | - | 2, 3 | 252 | |||
AK019381.1 | - | 2, 3 | 237 | |||
AK050647.1 | - | 2, 3 | 223 | |||
AK156743.1 | - | 222 | ||||
BC025796.1 | - | 2, 3 | 201 | 5100832 |