Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tmem25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027865.2 | NP_082141.1 | 7, 8 | 991 | ||
GenBank mRNA | AK002841.1 | - | 7, 8 | 992 | ||
AK090375.1 | - | 5, 6 | 1265 | |||
AK154206.1 | - | 6, 7 | 1477 | |||
AK165199.1 | - | 7, 8 | 1023 | |||
AK165827.1 | - | 7, 8 | 976 |