Search
Search

NDN| Gene Symbol: NDN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002487.2 | NP_002478.1 | 1 | 812 | ||
| GenBank mRNA | AK129654.1 | - | 2 | 710 | ||
| AK312779.1 | - | 1 | 806 | |||
| BC008750.1 | - | 1 | 789 | 3347128 | ||
| KJ534888.1 | - | 1 | 726 | |||
| KJ535010.1 | - | 2 | 326 | |||
| KU178221.1 | - | 1 | 726 | |||
| KU178222.1 | - | 2 | 528 | |||
| KU178223.1 | - | 2 | 404 | |||
| KU178224.1 | - | 2 | 310 | |||
| KU178225.1 | - | 2 | 243 | |||
| U35139.1 | - | 1 | 784 | |||