Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Lrrc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142728.1 | NP_001136200.1 | 21 | 3054 | ||
NM_030070.3 | NP_084346.2 | 21 | 3057 | |||
GenBank mRNA | AK019545.1 | - | 21 | 2990 | ||
AK029802.1 | - | 21 | 3055 |